COMH42116F87 Combi-Blocks, Inc.

Methyl 5-chloro-1-(2-methoxy-2-oxoethyl)-1h-1,2,4-triazole-3-carboxylate

  • MDL number MFCD31619035
  • Cas Number : 2279123-77-0
  • Molecular Weight : 233.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2790
Availability details for
Quantity
  • Availability
  • Add to Cart