COMH42116F88 Combi-Blocks, Inc.

N,N'-[1,4-Phenylenebis(oxyethane-2,1-diyl)]diacetamide

  • MDL number MFCD07658370
  • Cas Number : 893768-96-2
  • Molecular Weight : 280.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2805
Availability details for
Quantity
  • Availability
  • Add to Cart