COMH42116F92 Combi-Blocks, Inc.

Boc-l-met(o)-o-ch2-f-ch2-cooh dcha

  • MDL number MFCD30749174
  • Molecular Weight : 594.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2843

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2843
Availability details for
Quantity
  • Availability
  • Add to Cart