COMH42116F94 Combi-Blocks, Inc.

2-(2,5-Dimethoxyphenyl)-1-oxo-1,2-dihydroisoquinoline-4-carboxylic acid

  • MDL number MFCD24038561
  • Cas Number : 1429903-79-6
  • Molecular Weight : 325.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2849
Availability details for
Quantity
  • Availability
  • Add to Cart