COMH42116F99 Combi-Blocks, Inc.

1-Decanesulfonic acid sodium salt monohydrate

  • MDL number MFCD30491701
  • Molecular Weight : 262.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2857

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2857
Availability details for
Quantity
  • Availability
  • Add to Cart