COMH42116FAF Combi-Blocks, Inc.

1-2-[(2,4-Difluorophenyl)amino]-4-methylpyrimidin-5-ylethanone

  • MDL number MFCD31581341
  • Cas Number : 2270906-35-7
  • Molecular Weight : 263.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2946

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2946
Availability details for
Quantity
  • Availability
  • Add to Cart