COMH42116FB0 Combi-Blocks, Inc.

6-Hydroxy-2-(oxan-4-ylidene)-2,3-dihydro-1-benzofuran-3-one

  • MDL number MFCD30181226
  • Cas Number : 1955523-48-4
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-hydroxy-2-(tetrahydro-4H-pyran-4-ylidene)benzofuran-3(2H)-one
Inchi Code 1S/C13H12O4/c14-9-1-2-10-11(7-9)17-13(12(10)15)8-3-5-16-6-4-8/h1-2,7,14H,3-6H2
Inchi Key QAYXRCVLWHXECE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2947

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2947
Availability details for
Quantity
  • Availability
  • Add to Cart