COMH42116FB4 Combi-Blocks, Inc.

(Z)-6-Hydroxy-2-((1-methyl-1h-pyrrol-2-yl)methylene)benzofuran-3(2h)-one

  • MDL number MFCD28350038
  • Cas Number : 1630907-85-5
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-2980
Availability details for
Quantity
  • Availability
  • Add to Cart