COMH42116FCE Combi-Blocks, Inc.

3-Cyclopropyl-5-methyl-1h-pyrazole

  • MDL number MFCD20502732
  • Cas Number : 1287752-76-4
  • Molecular Weight : 122.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3113

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3113
Availability details for
Quantity
  • Availability
  • Add to Cart