COMH42116FD2 Combi-Blocks, Inc.

t-Boc-aminooxy-PEG1-amine

  • MDL number MFCD31561111
  • Cas Number : 1844894-82-1
  • Molecular Weight : 220.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3131

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3131
Availability details for
Quantity
  • Availability
  • Add to Cart