COMH42116FD7 Combi-Blocks, Inc.

1-Methyl-2-piperidin-2-yl-1h-benzimidazole dihydrochloride

  • MDL number MFCD31557150
  • Cas Number : 2270906-64-2
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3160

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3160
Availability details for
Quantity
  • Availability
  • Add to Cart