COMH42116FDF Combi-Blocks, Inc.

1,4,5,6,7,8-Hexahydrocyclohepta[c]pyrazole-3-carboxamide

  • MDL number MFCD27998369
  • Cas Number : 1823353-08-7
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3196

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3196
Availability details for
Quantity
  • Availability
  • Add to Cart