COMH42116FE1 Combi-Blocks, Inc.

[3-(Methoxymethyl)-1,2,4-oxadiazol-5-yl]methanol

  • MDL number MFCD18878788
  • Cas Number : 1344061-64-8
  • Molecular Weight : 144.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3199

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3199
Availability details for
Quantity
  • Availability
  • Add to Cart