COMH42116FE2 Combi-Blocks, Inc.

3-(Aminomethyl)-n,n-diethylpyridin-2-amine

  • MDL number MFCD09727760
  • Cas Number : 953738-54-0
  • Molecular Weight : 179.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(aminomethyl)-N,N-diethyl-2-pyridinamine
Inchi Code 1S/C10H17N3/c1-3-13(4-2)10-9(8-11)6-5-7-12-10/h5-7H,3-4,8,11H2,1-2H3
Inchi Key KWFAOZBFQTYAOE-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3204

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3204
Availability details for
Quantity
  • Availability
  • Add to Cart