COMH42116FE5 Combi-Blocks, Inc.

N-(2-Aminoethyl)-2-hydroxybenzamide hydrochloride

  • MDL number MFCD31583449
  • Cas Number : 92765-39-4
  • Molecular Weight : 216.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-aminoethyl)-2-hydroxybenzamide hydrochloride
Inchi Code 1S/C9H12N2O2.ClH/c10-5-6-11-9(13)7-3-1-2-4-8(7)12;/h1-4,12H,5-6,10H2,(H,11,13);1H
Inchi Key SNBOEMWUHFGLMN-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3211

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3211
Availability details for
Quantity
  • Availability
  • Add to Cart