COMH42116FE6 Combi-Blocks, Inc.

Tetrazine-sulfo-nhs ester

  • MDL number MFCD29052193
  • Cas Number : 1821017-48-4
  • Molecular Weight : 415.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3213

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3213
Availability details for
Quantity
  • Availability
  • Add to Cart