COMH42116FEB Combi-Blocks, Inc.

Methyl 2-methyl-3-(4-methylpiperidin-1-yl)propanoate

  • MDL number MFCD12193206
  • Cas Number : 23573-95-7
  • Molecular Weight : 199.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3241

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3241
Availability details for
Quantity
  • Availability
  • Add to Cart