COMH42116FF7 Combi-Blocks, Inc.

(3-Pyrrolidin-1-ylpropyl)amine dihydrochloride

  • MDL number MFCD27756346
  • Cas Number : 148736-33-8
  • Molecular Weight : 201.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3320

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3320
Availability details for
Quantity
  • Availability
  • Add to Cart