COMH42117002 Combi-Blocks, Inc.

(4-Piperidin-1-ylbutyl)amine dihydrobromide

  • MDL number MFCD12128983
  • Cas Number : 1158307-48-2
  • Molecular Weight : 318.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3391
Availability details for
Quantity
  • Availability
  • Add to Cart