COMH42117006 Combi-Blocks, Inc.

Tiamulin fumarate

  • MDL number MFCD27966802
  • Cas Number : 55297-96-6
  • Molecular Weight : 609.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3396

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3396
Availability details for
Quantity
  • Availability
  • Add to Cart