COMH4211700A Combi-Blocks, Inc.

Pc methyltetrazine-peg4-nhs carbonate ester

  • MDL number MFCD29052192
  • Cas Number : 2055736-28-0
  • Molecular Weight : 785.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3407

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3407
Availability details for
Quantity
  • Availability
  • Add to Cart