COMH4211700D Combi-Blocks, Inc.

N-[2-(3-Amino-4-methyl-phenoxy)-ethyl]-acetamide

  • MDL number MFCD22385559
  • Cas Number : 1394024-29-3
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[2-(3-amino-4-methylphenoxy)ethyl]acetamide
Inchi Code 1S/C11H16N2O2/c1-8-3-4-10(7-11(8)12)15-6-5-13-9(2)14/h3-4,7H,5-6,12H2,1-2H3,(H,13,14)
Inchi Key PLYHILUFTWSEQY-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3418

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3418
Availability details for
Quantity
  • Availability
  • Add to Cart