COMH4211700E Combi-Blocks, Inc.

7-Methoxy[1,2,4]triazolo[1,5-c]pyrimidin-5-amine

  • MDL number MFCD31583465
  • Cas Number : 2270906-79-9
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3422
Availability details for
Quantity
  • Availability
  • Add to Cart