COMH42117015 Combi-Blocks, Inc.

5-Cyclopropyl-1-methyl-1h-pyrazole-3-carboxamide

  • MDL number MFCD31557147
  • Cas Number : 1864712-18-4
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-cyclopropyl-1-methyl-1H-pyrazole-3-carboxamide
Inchi Code 1S/C8H11N3O/c1-11-7(5-2-3-5)4-6(10-11)8(9)12/h4-5H,2-3H2,1H3,(H2,9,12)
Inchi Key ROASBUWZSZGDBT-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3442
Availability details for
Quantity
  • Availability
  • Add to Cart