COMH42117019 Combi-Blocks, Inc.

1-(5-Bromo-1,3,4-thiadiazol-2-yl)piperidin-2-one

  • MDL number MFCD14281998
  • Cas Number : 1171433-64-9
  • Molecular Weight : 262.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(5-bromo-1,3,4-thiadiazol-2-yl)-2-piperidinone
Inchi Code 1S/C7H8BrN3OS/c8-6-9-10-7(13-6)11-4-2-1-3-5(11)12/h1-4H2
Inchi Key UBEDBLRJAAIQOV-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3466
Availability details for
Quantity
  • Availability
  • Add to Cart