COMH4211701D Combi-Blocks, Inc.

M-Peg4-(ch2)8-phosphonic acid ethyl ester

  • MDL number MFCD30458011
  • Cas Number : 2028281-89-0
  • Molecular Weight : 412.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3480

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3480
Availability details for
Quantity
  • Availability
  • Add to Cart