COMH4211701F Combi-Blocks, Inc.

[1-(3-Fluorobenzyl)-2,5-dioxoimidazolidin-4-yl]acetic acid

  • MDL number MFCD31557152
  • Cas Number : 2174896-12-7
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3483

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3483
Availability details for
Quantity
  • Availability
  • Add to Cart