COMH42117026 Combi-Blocks, Inc.

1,7-Dimethylpyrimido[4,5-d]pyrimidine-2,4(1h,3h)-dione

  • MDL number MFCD14716299
  • Cas Number : 1031577-44-2
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3513
Availability details for
Quantity
  • Availability
  • Add to Cart