COMH4211702E Combi-Blocks, Inc.

(2,3-Dimethyl-1-naphthyl)amine hydrochloride

  • MDL number MFCD31583459
  • Cas Number : 2323068-69-3
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dimethylnaphthalen-1-amine hydrochloride
Inchi Code 1S/C12H13N.ClH/c1-8-7-10-5-3-4-6-11(10)12(13)9(8)2;/h3-7H,13H2,1-2H3;1H
Inchi Key SFKGVJIVNINIKH-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3572
Availability details for
Quantity
  • Availability
  • Add to Cart