COMH42117031 Combi-Blocks, Inc.

3-(1-Oxo-1h-phthalazin-2-yl)-propionic acid

  • MDL number MFCD04119865
  • Cas Number : 53985-70-9
  • Molecular Weight : 218.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(1-oxo-2(1H)-phthalazinyl)propanoic acid
Inchi Code 1S/C11H10N2O3/c14-10(15)5-6-13-11(16)9-4-2-1-3-8(9)7-12-13/h1-4,7H,5-6H2,(H,14,15)
Inchi Key CNIZVVZEHJBRLJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3577

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3577
Availability details for
Quantity
  • Availability
  • Add to Cart