COMH42117032 Combi-Blocks, Inc.

1-[(4-Chloro-3,5-dimethylpyridin-2-yl)methyl]piperazine dihydrochloride

  • MDL number MFCD31557139
  • Cas Number : 2270906-09-5
  • Molecular Weight : 312.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3583
Availability details for
Quantity
  • Availability
  • Add to Cart