COMH42117034 Combi-Blocks, Inc.

T-Boc-n-amido-peg8-propargyl

  • MDL number MFCD31561113
  • Cas Number : 2183440-31-3
  • Molecular Weight : 507.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3586

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3586
Availability details for
Quantity
  • Availability
  • Add to Cart