COMH42117039 Combi-Blocks, Inc.

1,4-Dioxa-9-azaspiro[5.5]undecane

  • MDL number MFCD26710359
  • Cas Number : 855314-76-0
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3603

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3603
Availability details for
Quantity
  • Availability
  • Add to Cart