COMH42117044 Combi-Blocks, Inc.

8-Methoxy-4-methyl-1h-quinolin-2-one

  • MDL number MFCD02269198
  • Cas Number : 30198-01-7
  • Molecular Weight : 189.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3654
Availability details for
Quantity
  • Availability
  • Add to Cart