COMH42117046 Combi-Blocks, Inc.

Bis-peg2-endo-bcn

  • MDL number MFCD30527399
  • Cas Number : 1476737-97-9
  • Molecular Weight : 500.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3668

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3668
Availability details for
Quantity
  • Availability
  • Add to Cart