COMH42117047 Combi-Blocks, Inc.

N-(tert-Butyl)-1-phenylhydrazinecarbothioamide

  • MDL number MFCD31583443
  • Cas Number : 14294-28-1
  • Molecular Weight : 223.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(tert-butyl)-1-phenylhydrazine-1-carbothioamide
Inchi Code 1S/C11H17N3S/c1-11(2,3)13-10(15)14(12)9-7-5-4-6-8-9/h4-8H,12H2,1-3H3,(H,13,15)
Inchi Key FIAWPRWLVSPGLF-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3673

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3673
Availability details for
Quantity
  • Availability
  • Add to Cart