COMH42117054 Combi-Blocks, Inc.

[2-(1-Adamantyloxy)ethyl]methylamine

  • MDL number MFCD05854323
  • Cas Number : 21623-95-0
  • Molecular Weight : 209.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3714

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3714
Availability details for
Quantity
  • Availability
  • Add to Cart