COMH42117055 Combi-Blocks, Inc.

Pc mal-nhs carbonate ester

  • MDL number MFCD30458017
  • Cas Number : 1408057-91-9
  • Molecular Weight : 562.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3732

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3732
Availability details for
Quantity
  • Availability
  • Add to Cart