COMH42117056 Combi-Blocks, Inc.

(([2-(Methoxymethyl)-5-oxo-5h-[1,3,4]thiadiazolo[3,2-a]pyrimidin-7-yl]methyl)thio)acetic acid

  • MDL number MFCD16630637
  • Cas Number : 1239731-59-9
  • Molecular Weight : 301.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3735

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3735
Availability details for
Quantity
  • Availability
  • Add to Cart