COMH42117057 Combi-Blocks, Inc.

2-(2,4-Dioxo-1,4-dihydroquinazolin-3(2h)-yl)-3-phenylpropanoic acid

  • MDL number MFCD18488571
  • Molecular Weight : 310.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3737

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3737
Availability details for
Quantity
  • Availability
  • Add to Cart