COMH4211705F Combi-Blocks, Inc.

3-Aminobicyclo[2.2.1]hept-5-ene-2-carboxamide hydrochloride

  • MDL number MFCD27756494
  • Cas Number : 1983937-92-3
  • Molecular Weight : 188.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3783

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3783
Availability details for
Quantity
  • Availability
  • Add to Cart