COMH42117409 Combi-Blocks, Inc.

[1,2,4]Triazolo[1,5-a]pyridin-7-amine

  • MDL number MFCD22394733
  • Cas Number : 1396312-30-3
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Inchi Code 1S/C6H6N4/c7-5-1-2-10-6(3-5)8-4-9-10/h1-4H,7H2
Inchi Key MWDPRMZMHGSFSJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4195

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4195
Availability details for
Quantity
  • Availability
  • Add to Cart