COMH4211740B Combi-Blocks, Inc.

Methoxy peg maleimide

  • MDL number MFCD02688034
  • Cas Number : 99126-64-4
  • Molecular Weight : >5000

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5344

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-5344
Availability details for
Quantity
  • Availability
  • Add to Cart