COMH4211740D Combi-Blocks, Inc.

3-Nitropyrazolo[1,5-a]pyridine

  • MDL number MFCD28605492
  • Cas Number : 19363-97-4
  • Molecular Weight : 163.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7709

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-7709
Availability details for
Quantity
  • Availability
  • Add to Cart