COMH4211740E Combi-Blocks, Inc.

3-(4-Piperidyl)-[1,2,4]triazolo[4,3-a]pyridine hydrochloride

  • MDL number MFCD16496157
  • Cas Number : 1225150-46-8
  • Molecular Weight : 238.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3880
Availability details for
Quantity
  • Availability
  • Add to Cart