COMH42117411 Combi-Blocks, Inc.

3-(1-[(2-Methoxyphenyl)methyl]-2,5-dioxoimidazolidin-4-yl)propanoic acid

  • MDL number MFCD29051091
  • Cas Number : 1955492-68-8
  • Molecular Weight : 292.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3899

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3899
Availability details for
Quantity
  • Availability
  • Add to Cart