COMH42117419 Combi-Blocks, Inc.

2-(1-Benzyl-1h-pyrazol-5-yl)acetic acid

  • MDL number MFCD28530874
  • Cas Number : 1785465-63-5
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3934

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3934
Availability details for
Quantity
  • Availability
  • Add to Cart