COMH42117424 Combi-Blocks, Inc.

8,9-Difluoro-5,10-dihydrobenzo[5,6]thiepino[2,3-b]pyridin-5-ol

  • MDL number MFCD31630349
  • Molecular Weight : 265.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3975
Availability details for
Quantity
  • Availability
  • Add to Cart