COMH4211742A Combi-Blocks, Inc.

1-(Nitromethyl)cyclobutanol

  • MDL number MFCD21648486
  • Cas Number : 344329-87-9
  • Molecular Weight : 131.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-3998
Availability details for
Quantity
  • Availability
  • Add to Cart