COMH42117432 Combi-Blocks, Inc.

De-boc-docetaxel

  • MDL number MFCD30182048
  • Cas Number : 133524-69-3
  • Molecular Weight : 707.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HA-4021
Availability details for
Quantity
  • Availability
  • Add to Cart